Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ SED-CHEK 2™ ESR Controls
SDP

Catalog No. 2246919 Shop All Polymedco Products
Change view
Click to view available options
Type:
Abnormal Control
Bi-Level Control
Normal Control
For Use With (Equipment):
For Sedimat and Manual Procedures
For use with Sedimat™ 15 automated ESR analyzer
Sedimat™ 15 automated ESR analyzer
Quantity:
3 x 8 mL
6 x 8 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Quantity
22-469-19 Normal Control For use with Sedimat™ 15 automated ESR analyzer 6 x 8 mL
22-046912 Abnormal Control For Sedimat and Manual Procedures 6 x 8 mL
22-046913 Normal Control For Sedimat and Manual Procedures 6 x 8 mL
22-046911 Bi-Level Control For Sedimat and Manual Procedures 6 x 8 mL
22-469-17 Bi-Level Control Sedimat™ 15 automated ESR analyzer 3 x 8 mL
22-469-18 Abnormal Control For use with Sedimat™ 15 automated ESR analyzer 6 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-469-19 Supplier Polymedco™ Supplier No. ESR5CTN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unique cell-stabilizing process mimics the rouleaux formation of patient samples

  • Bi-level, whole-blood control designed to monitor ESR procedures
  • Monitor technique as well as environmental and physical factors that adversely affect ESR results
  • Run like a patient sample—not necessary to void diluent from pre-filled tubes
  • Universal control assayed for both automated and manual ESR methods

Order Info

Controls packaged in 6 x 8mL vials

Specifications

Cap Type Automatic self-zeroing
Compatible Methods Both automated and manual ESR methods
Content And Storage 20°C to 22°C
For Use With (Equipment) For use with Sedimat™ 15 automated ESR analyzer
Type Normal Control
Intended Use Whole Blood reference control material designed to monitor ESR procedures
Packaging Vial
Quantity 6 x 8 mL
Stability 31 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.