Learn More
Enzo Life Sciences Sedanolide (100mg). CAS: 6415-59-4
SDP
Supplier:  Enzo Life Sciences ALX350229M100
Encompass_Preferred
Inducer of glutathione S-transferases and inhibitor of chemically induced carcinogenesis. Mosquitocidal, nematicidal and antifungal. Was shown to inhibit cyclooxygenase-1 and -2 (COX-1 and COX-2) as well as topoisomerase I and topoisomerase II. Alternative name: 3-Butyl-3a,4,5,6-tetrahydro-1(3H)-isobenzofuranone. Purity: ≥98% (UHPLC). Solubility: Soluble in 100% ethanol or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-200-8788