Learn More
Electron Microscopy Sciences EMS Sedgewick-Rafter Counting Cell, Brass, 1.0 ml
SDP
Catalog No. 501928541
Encompass_Preferred
Intended for use with standard or inverted compound microscopes at low magnifications. Both cells include two cover glasses and one plastic case. Slide is 1" x 3". Cell size is 1.96" W x 0.787" H x 0.04" DCell volume is 1.0 ml.
Catalog No. 50-192-8541 Supplier Electron Microscopy Sciences Supplier No. 6350901
Intended for use with standard or inverted compound microscopes at low magnifications. Both cells include two cover glasses and one plastic case. Slide is 1" x 3". Cell size is 1.96" W x 0.787" H x 0.04" DCell volume is 1.0 ml.