Learn More
Electron Microscopy Sciences Sedwick-Rafter Gridded Counting Cell
SDP
Catalog No. 501928542
Encompass_Preferred
EMS Sedgewick-Rafter counting cells are intended for use with standard or inverted compound microscopes at low magnifications. These feature a millimeter square grid, providing the ability to extrapolate the number of zooplankton, thus saving time by not having to count the entire cell. Includes two cover glasses and one plastic case. Slide in 1 in x 3 in. Cell size is 1.96 in W x 0.787 in H x 0.04 in D. Cell volume is 1.0 ml.
Catalog No. 50-192-8542 Supplier Electron Microscopy Sciences Supplier No. 6350902
EMS Sedgewick-Rafter counting cells are intended for use with standard or inverted compound microscopes at low magnifications. These feature a millimeter square grid, providing the ability to extrapolate the number of zooplankton, thus saving time by not having to count the entire cell. Includes two cover glasses and one plastic case. Slide in 1 in x 3 in. Cell size is 1.96 in W x 0.787 in H x 0.04 in D. Cell volume is 1.0 ml.