Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ Sedimat™ 15 Plus Automated Sedrate System
SDP

Catalog No. 21000204 Shop All Polymedco Products
Change view
Click to view available options
Analysis Time:
15 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analysis Time
21-000-204 15 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-000-204 Supplier Polymedco™ Supplier No. SJ1215 PLUSBDL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Automated sedrate system that provides accurate ESR results in only 15 minute.

  • Fast, in-office results with random access
  • Can test one to eight specimens simultaneously
  • Fast throughput (process 32 patient results per hour) in a small footprint
  • Eight position reader provides computer interfacing to transfer data (LIS compatible)
  • Includes Sedimat 15 analyzer, base, printer and barcode reader

Specifications

Type Automated ESR Analyzer
Sample Type Blood
For Use With (Application) ESR testing
Analysis Time 15 min.
Throughput Process 32 patient results per hour
Includes Sedimat 15 analyzer, base, printer and barcode reader
Parameter ESR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.