Learn More
Polymedco™ Sedimat™ 15 Plus Sedrate System
SDP
Catalog No. 21000119 Shop All Polymedco Products
Change view
Click to view available options
Type:
Sedimat 15 Plus Base
Sedimat 15 Plus Sedrate System
Thermal Printer Paper
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21-000-119 Sedimat 15 Plus Base
13-641-587B Thermal Printer Paper
21-000-118 Sedimat 15 Plus Sedrate System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-000-119 Supplier Polymedco™ Supplier No. SJBASE PLUS
Add to Cart
Add to Cart
Fast, in-office results for one to eight specimens simultaneously.
Automated sedrate system provides accurate ESR results in only 15 minutes.
  • Random access capability
  • Faster throughput; processes 32 patient results per hour
  • Eight-position reader
  • Small footprint suitable for tight benchtops
  • Bar code reader available
  • LIS compatible for data transfer
  • Printer available (21-000-123)
Specifications
Type Sedimat 15 Plus Base
For Use With (Application) Fast, in-office results for one to eight specimens simultaneously
For Use With (Equipment) Sedimat™ 15 plus sedrate system