Learn More
Fisherfinest™ Sediplast™ Tubes With Diluent
SDP
Catalog No. 21000139 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 Tubes
Catalog No. Quantity
21-000-139 1000 Tubes
1 options
This item is not returnable. View return policy
Catalog No. 21-000-139
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Plastic tubes for optimum safety
  • Completely closed system
  • Self-zeroing cap and reservoir
  • Small sample volume requirement
  • Market-leading ESR tube
Specifications
Product Type Westergren Tube
Material Plastic
Sterility Non-sterile
For Use With (Equipment) Sediplast ESR system
Quantity 1000 Tubes
For Use With (Application) Diluent: Anticoagulant for whole blood samples