Learn More
Polymedco™ Sediten ESR Analyzer
SDP
Catalog No. 21000205 Shop All Polymedco Products
Change view
Click to view available options
Analysis Time:
10 min.
Catalog No. Analysis Time
21-000-205 10 min.
1 options
This item is not returnable. View return policy
Catalog No. 21-000-205 Supplier Polymedco™ Supplier No. 261091BDL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Analyzer, printer and barcode reader
An automated sedrate system that provides accurate ESR results in 10 minutes. Polymedco Sediten ESR Analyzer offers automation, convenience, improved workflow, speed and standardization of results.
  • Easy to operate
  • Small footprint
  • Random access, eight position reader
  • Fast throughput
  • Plastic tubes provides optimum safety
  • Tubes are available in vacuum and non-vacuum and with and without diluent
  • Completely closed system
  • Patient identifier tags available for barcode placement and/or sample ID
  • For laboratory setting
Specifications
Type Automated ESR Analyzer
Sample Type Blood
Analysis Time 10 min.
Throughput Fast
Includes Analyzer, printer and barcode reader
Parameter ESR