Learn More
Enzo Life Sciences SEEBRIGHT Gold 525 dUTP (25nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42843
Encompass_Preferred
Gold 525  [5(6)-Carboxyrhodamine 6G] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labeling, cDNA labeling and 3’-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Gold 525 dUTP can also be used for multicolor fluorescence labeling. Alternative name: 5(6)-Carboxyrhodamine 6G dUTP. Concentration: 1mM. Quantity: Sufficient for approximately 49 reactions, following the recommended protocol of ENZ-GEN111. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.
Catalog No. 50-201-2549