Learn More
Enzo Life Sciences SEEBRIGHT Mounting Medium (10ml)
SDP
Catalog No. 502013774
Encompass_Preferred
SEEBRIGHT Mounting Medium is an antifade reagent that minimizes photobleaching. Use of this mounting medium significantly extends the lifetime of fluorescent signals. Quantity: Each vial of SEEBRIGHT Mounting Medium contains 10 mL sufficient for 1000 tests (ENZ-GEN-420-1000). Apply mounting medium to the sample area as desired. Long Term Storage: +4°C. Applications: IF, Fluorescence microscopy, Fluorescent detection FISH.
Catalog No. 50-201-3774 Supplier Enzo Life Sciences Supplier No. ENZGEN4221000
SEEBRIGHT Mounting Medium is an antifade reagent that minimizes photobleaching. Use of this mounting medium significantly extends the lifetime of fluorescent signals. Quantity: Each vial of SEEBRIGHT Mounting Medium contains 10 mL sufficient for 1000 tests (ENZ-GEN-420-1000). Apply mounting medium to the sample area as desired. Long Term Storage: +4°C. Applications: IF, Fluorescence microscopy, Fluorescent detection FISH.