Learn More
Enzo Life Sciences SEEBRIGHT Red 650 dUTP (25nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42522
Encompass_Preferred
Red 650 [Cyanine-5E] dUTP is a cyanine-5  derivative with enhanced spectral properties. This nucleotide analog can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, cDNA labeling and 3'-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Red 650 dUTP can also be used for multicolor fluorescence labeling. Alternative name: Enhanced Cyanine 5-dUTP. Concentration: 1mM. Quantity: Sufficient for approximately 49 reactions, following the recommended protocol of ENZ-GEN111. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.
Catalog No. 50-201-2520