Learn More
Hampton Research Seed Bead Steel Kit, 24 tubes with Steel Seed Beads
SDP
Catalog No. NC1369311
Encompass
The Seed Bead kits are used to create a seed stock for performing subsequent seeding experiments. The Seed Bead kits contains 24 Seed Bead Tubes, a special 1.5 ml microcentrifuge tube, shaped to maintain the seed in the sample during mixing. Each tube is configured with a PTFE, Stainless Steel, Zirconium Silicate Ceramic, or Glass Seed Beads.
Catalog No. NC1369311 Supplier Hampton Research Supplier No. HR4780
The Seed Bead kits are used to create a seed stock for performing subsequent seeding experiments. The Seed Bead kits contains 24 Seed Bead Tubes, a special 1.5 ml microcentrifuge tube, shaped to maintain the seed in the sample during mixing. Each tube is configured with a PTFE, Stainless Steel, Zirconium Silicate Ceramic, or Glass Seed Beads.