Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Seed Germination and Plant Structure
GREENER_CHOICESCI_ED

Catalog No. S42854 Shop All Aldon Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42854 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING CHOKING HAZARD - Small parts. Not for children under 3 yrs.
Catalog No. S42854 Supplier Innovating Science™ Supplier No. IS1004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING CHOKING HAZARD - Small parts. Not for children under 3 yrs.

Innovating Science™ Seed Germination and Plant Structure allows young students to examine, and understand plant's needs for growth and survival

  • Allows students to dissect and germinate seeds, then compare their plant's growth to other types of plants grown in the classroom
  • Developed for grades K-2
  • Provides materials for 30 students (15 pairs)

Specifications

Color White
Grade K to 2
Age 5 to 7
Includes 15 Bean Seeds, 15 Magnifiers, 15 Toothpicks, 30 Plastic cups (5 oz.), 30 Pea seeds (7 oz.), 30 Sunflower seeds, 30 Wicks, Bag of soil
Quantity 1 Kit
Dimensions (L x W x H) 12.75 x 9.75 x 6.75 in.
Class Size 30 Students
Product Type Seed Germination and Plant Structure Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.