Learn More
Hampton Research Seeding Tool, 5 Pack
SDP
Catalog No. 500457
Encompass
Seeding Tool, 5 Pack Applications: Streak Seeding Features: Natural Fiber Built-In Handle/Cover The Seeding Tool Is A 1 Cm Natural Fiber Attached To A Stainless Steel Pin And Plastic Handle. The Seeding Tool Is Supplied With A Cover To Protect The Fiber And When The Cover Is Placed On The Back Side Of The Seeding Tool, It Makes An Excellent Handle.. Please note items from this supplier are non-returnable.
Catalog No. 50-045-7 Supplier Hampton Research Supplier No. HR8133
Seeding Tool, 5 Pack Applications: Streak Seeding Features: Natural Fiber Built-In Handle/Cover The Seeding Tool Is A 1 Cm Natural Fiber Attached To A Stainless Steel Pin And Plastic Handle. The Seeding Tool Is Supplied With A Cover To Protect The Fiber And When The Cover Is Placed On The Back Side Of The Seeding Tool, It Makes An Excellent Handle.. Please note items from this supplier are non-returnable.