Learn More
Medchemexpress LLC Sefaxersen sodium | 2272983-11-4 | 99.5% | 8678.96 (free acid) | 1 MG
SDP
Catalog No. 5000361312
Encompass_Preferred
Sefaxersen sodium is a specific antisense oligonucleotide (ASO) targeting complement factor B (CFB). It effectively reduces circulating levels of CFB and can be used for geographic atrophy (GA) research.
  • Specific antisense oligonucleotide (ASO)
  • Targets complement factor B (CFB)
  • Reduces circulating levels of CFB
  • Used for geographic atrophy (GA) research
Catalog No. 50-003-61312 Supplier Medchemexpress LLC Supplier No. HY148370A1MG
Sefaxersen sodium is a specific antisense oligonucleotide (ASO) targeting complement factor B (CFB). It effectively reduces circulating levels of CFB and can be used for geographic atrophy (GA) research.
  • Specific antisense oligonucleotide (ASO)
  • Targets complement factor B (CFB)
  • Reduces circulating levels of CFB
  • Used for geographic atrophy (GA) research