Learn More
Selleck Chemical LLC Segesterone acetate-E0937-5MG
SDP
Catalog No. 503137167
Encompass_Preferred
Segesterone acetate (Nestoron ST-1435) is a synthetic progestin that binds to the progesterone receptor with EC50 of 50 3 nM
Catalog No. 50-313-7167 Supplier Selleck Chemical LLC Supplier No. E09375MG
Segesterone acetate (Nestoron ST-1435) is a synthetic progestin that binds to the progesterone receptor with EC50 of 50 3 nM