Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Segetalin A | 161875-97-4 | 99.9% | 609.72 | C31H43N7O6 | 1 MG
SDP

Catalog No. 5000240434
Encompass_Preferred

Segetalin A is a cyclic hexapeptide isolated from the seeds of Vaccaria hispanica, provided as a research-grade reference compound for biochemical and pharmacological studies.

  • Compound isolated from Vaccaria hispanica.
  • CAS 161875-97-4.
  • Molecular formula C31H43N7O6; molecular weight 609.72.
  • Purity 99.92%.
  • White to off-white solid for laboratory use.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • COA and SDS available.
  • For research use only; not for human use.

Catalog No. 50-002-40434 Supplier Medchemexpress LLC Supplier No. HY1072461MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Segetalin A is a cyclic hexapeptide isolated from the seeds of Vaccaria hispanica, provided as a research-grade reference compound for biochemical and pharmacological studies.

  • Compound isolated from Vaccaria hispanica.
  • CAS 161875-97-4.
  • Molecular formula C31H43N7O6; molecular weight 609.72.
  • Purity 99.92%.
  • White to off-white solid for laboratory use.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • COA and SDS available.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.