Learn More
Medchemexpress LLC Seglitide acetate 10mg | 99248-33-6 | 10MG
SDP
Catalog No. 5000244397
Encompass_Preferred
Seglitide acetate (MK 678 L 36358 acetate) is a potent orally active somatostatin receptor 2 (SSTR2) agonist and also a competitive antagonist of SSTR14 SSTR25 and SSTR28 Seglitide acetate has antihypertensive effects and can inhibit plasma glucagon and growth hormone Seglitide acetate can be used for research on diabetes[1 [2 [3
Catalog No. 50-002-44397 Supplier Medchemexpress LLC Supplier No. HY106103A10MG
Seglitide acetate (MK 678 L 36358 acetate) is a potent orally active somatostatin receptor 2 (SSTR2) agonist and also a competitive antagonist of SSTR14 SSTR25 and SSTR28 Seglitide acetate has antihypertensive effects and can inhibit plasma glucagon and growth hormone Seglitide acetate can be used for research on diabetes[1 [2 [3