Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Seglitide acetate | 99248-33-6 | 99.3% | 869.02 g/mol | C46H60N8O9 | 1 MG
SDP

Catalog No. 5000243598
Encompass_Preferred

Seglitide acetate is a cyclic somatostatin analog peptide used as a research reagent to probe somatostatin receptor pharmacology, including SSTR2-mediated signaling and metabolic effects. Provided as the acetate salt in small, research-scale quantities, it is intended for laboratory use only.

  • Somatostatin receptor 2 (SSTR2) agonist for receptor pharmacology studies.
  • Cyclic peptide provided as the acetate salt.
  • High purity: 99.3% (HPLC).
  • Molecular weight: 869.02 g/mol; formula C46H60N8O9.
  • Supplied in research sizes (1 mg-100 mg).
  • Accompanying analytical documentation: datasheet, COA, RP-HPLC, MS, elemental analysis, SDS.

Catalog No. 50-002-43598 Supplier Medchemexpress LLC Supplier No. HY106103A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seglitide acetate is a cyclic somatostatin analog peptide used as a research reagent to probe somatostatin receptor pharmacology, including SSTR2-mediated signaling and metabolic effects. Provided as the acetate salt in small, research-scale quantities, it is intended for laboratory use only.

  • Somatostatin receptor 2 (SSTR2) agonist for receptor pharmacology studies.
  • Cyclic peptide provided as the acetate salt.
  • High purity: 99.3% (HPLC).
  • Molecular weight: 869.02 g/mol; formula C46H60N8O9.
  • Supplied in research sizes (1 mg-100 mg).
  • Accompanying analytical documentation: datasheet, COA, RP-HPLC, MS, elemental analysis, SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.