Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Seglitide acetate | 99248-33-6 | 99.3% | 869.02 g/mol | C46H60N8O9 | 5 MG
SDP

Catalog No. 5000244431
Encompass_Preferred

Seglitide acetate is a cyclic peptide somatostatin receptor 2 (SSTR2) agonist used in pharmacology research to study somatostatin receptor signaling and regulation of hormone secretion. Supplied as an acetate salt in solid form, it is typically dissolved in DMSO for in vitro and in vivo experiments probing glucagon and growth hormone modulation.

  • Potent SSTR2 agonist activity.
  • Cyclic peptide sequence for receptor specificity.
  • High purity, 99.3%.
  • Available in small milligram quantities for research use.
  • Soluble in DMSO for straightforward sample preparation.

Catalog No. 50-002-44431 Supplier Medchemexpress LLC Supplier No. HY106103A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seglitide acetate is a cyclic peptide somatostatin receptor 2 (SSTR2) agonist used in pharmacology research to study somatostatin receptor signaling and regulation of hormone secretion. Supplied as an acetate salt in solid form, it is typically dissolved in DMSO for in vitro and in vivo experiments probing glucagon and growth hormone modulation.

  • Potent SSTR2 agonist activity.
  • Cyclic peptide sequence for receptor specificity.
  • High purity, 99.3%.
  • Available in small milligram quantities for research use.
  • Soluble in DMSO for straightforward sample preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.