Learn More
Medchemexpress LLC sEH inhibitor-1 | 1208549-68-1 | 99.6% | 1 MG

Supplier: Medchemexpress LLC HY1204941MG
sEH inhibitor-1 (compound TCPU) is a research-grade, orally active inhibitor of soluble epoxide hydrolase (sEH) supplied as a powder for biochemical and preclinical studies. It shows sub-nanomolar to low-nanomolar potency in enzymatic assays and is intended for in vitro and in vivo research.
- Potent sEH inhibition: human IC50 0.4 nM, murine IC50 5.3 nM.
- High purity, typically 99.6%.
- Molecular weight 371.35; formula C17H20F3N3O3.
- Stable as powder at -20°C for long-term storage.
- Suitable for in vitro enzymatic assays and in vivo preclinical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.