Learn More
Medchemexpress LLC sEH inhibitor-1 (compound TCPU) | 1208549-68-1 | 99.6% | 10 MG
SDP
Catalog No. 5000234397
Encompass_Preferred
sEH inhibitor-1 (TCPU) is a potent, orally active inhibitor of soluble epoxide hydrolase (sEH) with low-nanomolar potency in human and murine assays. It is supplied as a high-purity research-grade compound for biochemical and pharmacological studies.
  • Low-nanomolar potency (human IC50 0.4 nM; murine IC50 5.3 nM).
  • Molecular formula C17H20F3N3O3 and molecular weight 371.35 g/mol.
  • High purity suitable for research (99.62%).
  • Available in small milligram pack sizes for in vitro and in vivo experiments.
  • Targets soluble epoxide hydrolase for mechanistic and pharmacological studies.
Catalog No. 50-002-34397 Supplier Medchemexpress LLC Supplier No. HY12049410MG
sEH inhibitor-1 (TCPU) is a potent, orally active inhibitor of soluble epoxide hydrolase (sEH) with low-nanomolar potency in human and murine assays. It is supplied as a high-purity research-grade compound for biochemical and pharmacological studies.
  • Low-nanomolar potency (human IC50 0.4 nM; murine IC50 5.3 nM).
  • Molecular formula C17H20F3N3O3 and molecular weight 371.35 g/mol.
  • High purity suitable for research (99.62%).
  • Available in small milligram pack sizes for in vitro and in vivo experiments.
  • Targets soluble epoxide hydrolase for mechanistic and pharmacological studies.