Learn More
Medchemexpress LLC sEH inhibitor-1 (compound TCPU) | 1208549-68-1 | 99.6% | 25 MG
SDP
Catalog No. 5000234544
Encompass_Preferred
sEH inhibitor-1 (compound TCPU) is a potent, orally active soluble epoxide hydrolase (sEH) inhibitor intended for research. It shows IC50 values of 0.4 nM in human sEH and 5.3 nM in murine sEH, and is supplied as a high-purity solid with strong DMSO solubility.
  • Potent sEH inhibition (IC50 0.4 nM human; 5.3 nM murine).
  • High purity (99.6%).
  • Molecular formula C17H20F3N3O3; molecular weight 371.35 g/mol.
  • Good solubility in DMSO (125 mg/mL).
  • Stable when stored under recommended conditions.
Catalog No. 50-002-34544 Supplier Medchemexpress LLC Supplier No. HY12049425MG
sEH inhibitor-1 (compound TCPU) is a potent, orally active soluble epoxide hydrolase (sEH) inhibitor intended for research. It shows IC50 values of 0.4 nM in human sEH and 5.3 nM in murine sEH, and is supplied as a high-purity solid with strong DMSO solubility.
  • Potent sEH inhibition (IC50 0.4 nM human; 5.3 nM murine).
  • High purity (99.6%).
  • Molecular formula C17H20F3N3O3; molecular weight 371.35 g/mol.
  • Good solubility in DMSO (125 mg/mL).
  • Stable when stored under recommended conditions.