Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC sEH inhibitor-1 (compound TCPU) | 1208549-68-1 | 99.6% | 50 MG
SDP

Catalog No. 5000232896
Encompass_Preferred

sEH inhibitor-1 (TCPU) is a potent, orally active inhibitor of soluble epoxide hydrolase (sEH) used in preclinical research. It exhibits sub-nanomolar to low-nanomolar potency against human and murine sEH and is supplied as a research-grade powder.

  • Potent sEH inhibition: IC50 0.4 nM (human), 5.3 nM (murine).
  • Chemical formula C17H20F3N3O3, molecular weight 371.35 g/mol.
  • High purity (~99.6%) suitable for research use.
  • Supplied as a powder; recommended storage at -20°C for long-term, 4°C for shorter-term.
  • Used in studies of inflammation, cardiovascular disease, and metabolic disorders.
  • Available in a 50 mg pack size.

Catalog No. 50-002-32896 Supplier Medchemexpress LLC Supplier No. HY12049450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

sEH inhibitor-1 (TCPU) is a potent, orally active inhibitor of soluble epoxide hydrolase (sEH) used in preclinical research. It exhibits sub-nanomolar to low-nanomolar potency against human and murine sEH and is supplied as a research-grade powder.

  • Potent sEH inhibition: IC50 0.4 nM (human), 5.3 nM (murine).
  • Chemical formula C17H20F3N3O3, molecular weight 371.35 g/mol.
  • High purity (~99.6%) suitable for research use.
  • Supplied as a powder; recommended storage at -20°C for long-term, 4°C for shorter-term.
  • Used in studies of inflammation, cardiovascular disease, and metabolic disorders.
  • Available in a 50 mg pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.