Learn More
Medchemexpress LLC sEH inhibitor-7 | 340221-20-7 | 99.5% | 247.33 | 100 MG
SDP
Catalog No. 5000361626
Encompass_Preferred
sEH inhibitor-7 is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH). Soluble epoxide hydrolase plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
  • Inhibits soluble epoxide hydrolase (sEH)
  • IC50s of 0.15 μM (mouse sEH)
  • IC50s of 6.2 μM (human sEH)
  • Plays a role in the metabolism of endogenous chemical mediators
  • Involved in blood pressure regulation
  • Involved in inflammation regulation
  • For research use only
Catalog No. 50-003-61626 Supplier Medchemexpress LLC Supplier No. HY148136100MG
sEH inhibitor-7 is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH). Soluble epoxide hydrolase plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
  • Inhibits soluble epoxide hydrolase (sEH)
  • IC50s of 0.15 μM (mouse sEH)
  • IC50s of 6.2 μM (human sEH)
  • Plays a role in the metabolism of endogenous chemical mediators
  • Involved in blood pressure regulation
  • Involved in inflammation regulation
  • For research use only