Learn More
Medchemexpress LLC Seh inhibitor-7 | 340221-20-7 | >99.5% | 247.33 | 50 MG
SDP
Catalog No. 5000363036
Encompass_Preferred
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH), respectively. Soluble epoxide hydrolase (sEH) plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
  • Inhibitor of soluble epoxide hydrolase (sEH).
  • Involved in the regulation of blood pressure.
  • Involved in the regulation of inflammation.
Catalog No. 50-003-63036 Supplier Medchemexpress LLC Supplier No. HY14813650MG
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH), respectively. Soluble epoxide hydrolase (sEH) plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
  • Inhibitor of soluble epoxide hydrolase (sEH).
  • Involved in the regulation of blood pressure.
  • Involved in the regulation of inflammation.