Learn More
Medchemexpress LLC sEH inhibitor-7 | 340221-20-7 | 99.5% | 247.33 g/mol | C15H21NO2 | 10 MG
SDP
Catalog No. 5000286041
Encompass_Preferred
sEH inhibitor-7 is a small-molecule research compound that inhibits soluble epoxide hydrolase (sEH). It is intended for in vitro and preclinical studies and is supplied as a solid or as a 10 mM solution in DMSO. Key properties, activity data, and handling recommendations are provided in the product datasheet and SDS.
  • Inhibits soluble epoxide hydrolase with IC50 = 0.15 μM (mouse) and 6.2 μM (human).
  • High purity: 99.5%.
  • Molecular weight 247.33 g/mol; formula C15H21NO2.
  • Available as solid and 10 mM solution in DMSO, including 10 mg pack sizes.
  • Powder stable at -20°C for up to 3 years; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Supplied with SDS and datasheet for safety and handling information.
Catalog No. 50-002-86041 Supplier Medchemexpress LLC Supplier No. HY14813610MG
sEH inhibitor-7 is a small-molecule research compound that inhibits soluble epoxide hydrolase (sEH). It is intended for in vitro and preclinical studies and is supplied as a solid or as a 10 mM solution in DMSO. Key properties, activity data, and handling recommendations are provided in the product datasheet and SDS.
  • Inhibits soluble epoxide hydrolase with IC50 = 0.15 μM (mouse) and 6.2 μM (human).
  • High purity: 99.5%.
  • Molecular weight 247.33 g/mol; formula C15H21NO2.
  • Available as solid and 10 mM solution in DMSO, including 10 mg pack sizes.
  • Powder stable at -20°C for up to 3 years; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Supplied with SDS and datasheet for safety and handling information.