Learn More
Medchemexpress LLC Seh inhibitor-7 | 340221-20-7 | 99.50% | 247.33 | 5 MG
SDP
Catalog No. 5000363682
Encompass_Preferred
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH), respectively. Soluble epoxide hydrolase (sEH) plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
  • Inhibitor of soluble epoxide hydrolase (sEH)
  • IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH)
  • Plays an important role in the metabolism of endogenous chemical mediators
  • Involved in the regulation of blood pressure and inflammation
Catalog No. 50-003-63682 Supplier Medchemexpress LLC Supplier No. HY1481365MG
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH), respectively. Soluble epoxide hydrolase (sEH) plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
  • Inhibitor of soluble epoxide hydrolase (sEH)
  • IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH)
  • Plays an important role in the metabolism of endogenous chemical mediators
  • Involved in the regulation of blood pressure and inflammation