Learn More
Enterprise Technology Solutions Seiko Address Labels, 1-1/8"x3-1/2", 130/Roll, 2/Each, White
SDP
Catalog No. 5000739777
Encompass_Preferred
High-quality address labels streamline your labeling process and ensure crisp, legible printouts. Strong adhesive securely sticks to your envelopes. Address labels are designed for Seiko SLP 400, 200 and 100 series, SLP EZ30 and Dymo Labelwriter 300 and 400 series printers.
Catalog No. 50-007-39777 Supplier Enterprise Technology Solutions Supplier No. SKPSLP2RL
High-quality address labels streamline your labeling process and ensure crisp, legible printouts. Strong adhesive securely sticks to your envelopes. Address labels are designed for Seiko SLP 400, 200 and 100 series, SLP EZ30 and Dymo Labelwriter 300 and 400 series printers.