Learn More
AdipoGen Sekikaic acid (1 mg)
SDP
Supplier:  AdipoGen AGCN20502M001
Encompass_Preferred
Chemical. CAS: 607-11-4. Formula: C22H26O8. MW: 418.4. Isolated from Ramalina sp. Antioxidant. Reactive oxygen species (ROS) scavenger. Specific ligand of the coactivator CBP/p300, binding to the GACKIX domain. Efficiently inhibits binding of activators at two binding sites. Antibiotic. Potent antibacterial compound. Antifungal on selected fungi species. Antiviral agent. Interferes with the viral replication of selected respiratory syncytial virus. Competitive alpha-glucosidase and non-competitive beta-glucosidase inhibitor. Shows moderate anticancer activity.
Catalog No. 50-205-3685