Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ hCG Card Pregnancy Test Kit
GSA_VA

Catalog No. 22071064 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Format:
Card
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-071-064 Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-071-064 Supplier SEKISUI Diagnostics™ Supplier No. 102W

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The OSOM™ hCG Card Pregnancy Test Kit is a simple, easy-to-read cassette-style immunoassay test for detection of hCG in urine for early determination of pregnancy.

  • Simple-to-use - just one step
  • Easy-to-read, black band(s) on white results
  • Trust your results - no biotin interference and detection limits as low as 20 mIU/mL
  • Get results quickly - results in three minutes
  • Ready to use - includes transfer pipette for each test

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
CPT Code 81025QW
Clia Complexity Waived
Detectable Analytes hCG beta-core Fragments
Format Card
Sample Type Urine
Sensitivity 20 mIU/mL
Type Rapid Test
Includes 25 card pregnancy test devices individually pouched, each containing a disposable pipette, instructions for use (IFU)
Packaging Type Test Kit
Test Time 3 min.
Analysis Time 3 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.