Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ hCG Combo Test

Catalog No. 22071060 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Format:
Cassette, Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-071-060 Cassette, Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-071-060 Supplier SEKISUI Diagnostics™ Supplier No. 124

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The OSOM™ hCG Combo Test is a simple, easy-to-read cassette-style immunoassay test for detection of hCG in urine or serum for the early detection of pregnancy.

  • hCG Combo Urine and Serum Test Kit
  • Simple-to-use - just one step
  • Easy-to-read, black band(s) on white results
  • Trust your results - no biotin interference and detection limits as low as 10 mIU/mL for serum, and 20 mIU/mL for urine
  • Get results quickly - results in three minutes (urine) and five minutes (serum)
  • Ready to use - includes transfer pipette for each test
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
CPT Code 81025QW (Urine), 84703 (Serum)
Clia Complexity Waived (Urine), Non-waived (Serum)
Detectable Analytes hCG beta-core Fragments
Format Cassette, Kit
Sample Type Serum, Urine
Sensitivity 10 mIU/mL Serum, 20 mIU/mL Urine
Type Rapid Test
Includes 25 combo test devices individually pouched, each containing a disposable pipette, Instructions for use (IFU), Pictorial procedure for convenient reference
Packaging Type Test Kit
Test Time 3 min. (Urine), 5 min. (Serum)
Analysis Time 3 to 5 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.