Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ hCG Urine Test Kit
GSA_VA

Catalog No. 22071063 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Format:
Manual Dipstick
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-071-063 Manual Dipstick
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-071-063 Supplier SEKISUI Diagnostics™ Supplier No. 101

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

OSOM™ hCG Urine Test Kit offers an economical, easy-to-read dipstick for the qualitative determination of hCG in urine.

  • Simple-to-use - just one step
  • Easy-to-read, two-color results
  • Get results quickly - results in as little as one minute, with confirmation of negatives at five minutes
  • Trust your results - no biotin interference and detection limits as low as 25 mIU/mL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
CPT Code 81025QW
Clia Complexity Waived
Detectable Analytes hCG beta-core Fragments
Format Manual Dipstick
Sample Type Urine
Sensitivity 25 mIU/mL
Type Rapid Test
Includes Container of 50 test sticks and instructions for use (IFU)
Packaging Type Test Kit
Test Time 5 min.
Analysis Time 5 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.