Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ Ultra Flu A and B Rapid Test

Catalog No. 22321105 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Type:
OSOM Ultra Flu A and B Nasopharyngeal Wash/Aspirate Kit
OSOM Ultra Flu A and B Rapid Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-321-105 OSOM Ultra Flu A and B Rapid Test
22-321-106 OSOM Ultra Flu A and B Nasopharyngeal Wash/Aspirate Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-321-105 Supplier SEKISUI Diagnostics™ Supplier No. 1006

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

OSOM™ Ultra Flu A and B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.

Improved Outcomes: Rapid and accurate diagnosis of Influenza enables a choice of antiviral therapy, helps avoid the overuse of antibiotics, and prevents healthcare costs related to unnecessary testing and treatment.

Patient Experience: Streamlines lab's workflow while reducing patient wait time and unnecessary use of antibiotics.

  • For the detection of Influenza type A and type B nucleoprotein antigens
  • Able to meet FDA reclassification standards
  • Multiple specimen types: nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
  • Cleared for use with multiple viral transport media (VTM) types
  • Accuracy: Influenza A Sensitivity: >90%; Influenza B: >85% vs culture (across multiple sample types)
  • Fast: Obtain positive results in 10 to 15 minutes
  • Flexible: Runs from nasal swab, NP swab as well as nasal aspirate/wash
  • Workflow: approved for use with multiple viral transport media types
  • QC Inside: 2 extra no charge cassettes to the kit

OSOM Ultra Flu A and B Control Kit (Cat. No. 22321114): 5 x positive Flu A/B swabs and 5 x Negative Flu A/B swabs
OSOM Ultra Flu A and B Nasopharyngeal Wash/Aspirate Kit (Cat. No. 22321106): 50 pipets and 5 mL extraction reagent
OSOM Ultra Flu A and B Swab Transport Kit (Cat. No. 22321107): 25 swabs in transport tubes

Storage Requirements:Store at room temperature.

Compliance

Meets FDA Reclassification

Recommended Storage

Store at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Cleared for use with multiple viral transport media (VTM) types
Content And Storage Room Temperature
CPT Code Flu A: 87804; Flu B: 87804-59 (modifier used to indicate separate test)
Clia Complexity Waived, Moderate
Detectable Analytes Influenza A and B
Format Kit
Sample Type Nasal and Nasopharyngeal Swab, Nasal Aspirate/Wash
Sensitivity Influenza A: 89.2%, Influenza B: 86.4% vs. PCR
Type OSOM Ultra Flu A and B Rapid Test
Includes 25 Test Kits
Specificity Influenza A: 99.4%, Influenza B: 99% vs PCR
Test Time 10 to 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.