Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ Swabs for Rapid Microbial Testing

Catalog No. 22071077 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Format:
Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-071-077 Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-071-077 Supplier SEKISUI Diagnostics™ Supplier No. 7628
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Swabs for Rapid Microbial Testing for use with Microbiological Specimens.

For use with Microbiological Specimen

  • For single use only
  • Made of Rayon
TRUSTED_SUSTAINABILITY

Specifications

Product Type Rayon Swabs
Format Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.