Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ H. pylori Test

Catalog No. 22071201 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Type:
Control Kit
Rapid Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-071-201 Rapid Test
22-071-080 Control Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-071-201 Supplier SEKISUI Diagnostics™ Supplier No. 175
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Use the OSOM™ H. pylori Test to detect Anti-H. pylori IgG antibodies in just three steps and 10 minutes.

  • Simple-to-use - get easy to read results with one reagent and three easy steps, with no need for additional equipment
  • Minimal sample required - test requires 10 μL of serum or plasma; 25 μL of whole blood
  • Trust your results - sensitivity of 95.9% versus biopsy/histology
  • Easy-to-read - two colored lines at both the test (T) and control (C) positions indicate a positive result
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature (2°C to 30°C, 35.6°F to 86°F)
Control Sets Not included
CPT Code 86318QW (Whole Blood), 86677 (Serum, Plasma)
CE Marker Yes
Clia Complexity Waived (Whole Blood)
Detectable Analytes H. pylori
Format Cassette
Sample Type Whole Blood, Serum, or Plasma
Sensitivity 95.9%
Type Rapid Test
Includes 25 test devices, 25 capillary tubes, developer solution, instructions for use (IFU)
Specificity 89.1%
Test Time 10 min.
Analysis Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.