Learn More
Description
- For use with OSOM™ Trichomonas and OSOM™ BVBLUE™ test kits
- 50 sets per box
Specifications
Specifications
| Tip Material | Cotton |
| Content And Storage | Room Temperature |
| For Use With (Application) | OSOM BVBLUE and Trichomonas Tests |
| No. per Pack | 50 Pk. |
| Product Type | Swab |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.