Learn More
Selleck Chemical LLC SEL120 S8840-25MG
SDP
Catalog No. 5000025162
Encompass_Preferred
SEL120 (SEL120-34 SEL120-34A) is a novel inhibitor of Cyclin-dependent kinase 8 (CDK8) with IC50 values of 4 4 nM and 10 4 nM for CDK8/Cyclin C and CDK19/CyclinC respectively
Catalog No. 50-000-25162 Supplier Selleck Chemical LLC Supplier No. S884025MG
SEL120 (SEL120-34 SEL120-34A) is a novel inhibitor of Cyclin-dependent kinase 8 (CDK8) with IC50 values of 4 4 nM and 10 4 nM for CDK8/Cyclin C and CDK19/CyclinC respectively