Learn More
Selleck Chemical LLC SEL120 (SEL120-34A) hydrochloride 1mg 1609522-33-9 SEL120-34
SDP
Supplier:  Selleck Chemical LLC S88401MG
Encompass_Preferred
SEL120 (SEL120-34, SEL120-34A) is a novel inhibitor of Cyclin-dependent kinase 8 (CDK8) with IC50 values of 4.4 nM and 10.4 nM for CDK8/Cyclin C and CDK19/CyclinC respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1706428