Learn More
Cayman Chemical SEL120-34A (hydrochloride), Purity: ≥98%, CAS: 1609452-30-3, 10 mg
SDP
Catalog No. 502941205
Encompass_Preferred
A dual inhibitor of Cdk8 and Cdk19 (IC50s = 4.4 and 10.4 nM, respectively); selective for Cdk8 and Cdk19 over a panel of CDK/cyclin complexes at 1 µM; inhibits IFN-γ-induced phosphorylation of STAT1 in HCT116 colon cancer cells at 0.1-5 µM; inhibits proliferation of MOLM-16, MV4-11, and KG-1 cells (GI50s = <1 µM); reduces tumor growth in a KG-1 mouse xenograft model at 60 mg/kg
Catalog No. 50-294-1205 Supplier Cayman Chemical Supplier No. 2790610 MG
A dual inhibitor of Cdk8 and Cdk19 (IC50s = 4.4 and 10.4 nM, respectively); selective for Cdk8 and Cdk19 over a panel of CDK/cyclin complexes at 1 µM; inhibits IFN-γ-induced phosphorylation of STAT1 in HCT116 colon cancer cells at 0.1-5 µM; inhibits proliferation of MOLM-16, MV4-11, and KG-1 cells (GI50s = <1 µM); reduces tumor growth in a KG-1 mouse xenograft model at 60 mg/kg