Learn More
Medchemexpress LLC Selachyl alcohol | 593-31-7 | C21H42O3 | 1 ML
SDP
Catalog No. 5000372032
Encompass_Preferred
Selachyl alcohol is an orally active antihypertensive agent that exhibits similar activities to antihypertensive neutral renomedullary lipid (ANRL). It is an alkylglycerol compound found in shark liver oil mixtures and has properties that can reduce lung metastasis. This compound can be used for cardiovascular disease research.
  • Orally active antihypertensive agent
  • Reduces lung metastasis
  • Can be used for cardiovascular disease research
  • Reduces blood pressure in hypertensive models
Catalog No. 50-003-72032 Supplier Medchemexpress LLC Supplier No. HY15289910MM/1ML
Selachyl alcohol is an orally active antihypertensive agent that exhibits similar activities to antihypertensive neutral renomedullary lipid (ANRL). It is an alkylglycerol compound found in shark liver oil mixtures and has properties that can reduce lung metastasis. This compound can be used for cardiovascular disease research.
  • Orally active antihypertensive agent
  • Reduces lung metastasis
  • Can be used for cardiovascular disease research
  • Reduces blood pressure in hypertensive models