Learn More
Medchemexpress LLC Selachyl alcohol | 593-31-7 | C21H42O3 | 5 MG
SDP
Catalog No. 5000374084
Encompass_Preferred
Selachyl alcohol is an orally active antihypertensive agent with activities similar to antihypertensive neutral renomedullary lipid (ANRL). It is an alkylglycerol compound found in shark liver oil, possessing properties that reduce lung metastasis. It can be used for cardiovascular disease research.
  • Reduces blood pressure in hypertensive mice.
  • Induces a slow drop in blood pressure of approximately 60 mm Hg.
  • Effectively reduces blood pressure via the gastrointestinal tract.
Catalog No. 50-003-74084 Supplier Medchemexpress LLC Supplier No. HY1528995MG
Selachyl alcohol is an orally active antihypertensive agent with activities similar to antihypertensive neutral renomedullary lipid (ANRL). It is an alkylglycerol compound found in shark liver oil, possessing properties that reduce lung metastasis. It can be used for cardiovascular disease research.
  • Reduces blood pressure in hypertensive mice.
  • Induces a slow drop in blood pressure of approximately 60 mm Hg.
  • Effectively reduces blood pressure via the gastrointestinal tract.