Learn More
Medchemexpress LLC Seladelpar (lysine) | 928821-41-4 | 98.0 % | 590.65 | 5 MG
SDP
Catalog No. 5000401792
Encompass_Preferred
Seladelpar lysine is an orally active, potent, and specific PPARδ agonist with an EC50 of 2 nM. It exhibits more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors, respectively. Seladelpar lysine can be used for the study of primary biliary cholangitis.
  • Orally active, potent, and specific PPARδ agonist
  • High selectivity over PPARα and PPARγ receptors (>750-fold and >2500-fold, respectively)
  • Can be used for the study of primary biliary cholangitis
Catalog No. 50-004-01792 Supplier Medchemexpress LLC Supplier No. HY19522B5MG
Seladelpar lysine is an orally active, potent, and specific PPARδ agonist with an EC50 of 2 nM. It exhibits more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors, respectively. Seladelpar lysine can be used for the study of primary biliary cholangitis.
  • Orally active, potent, and specific PPARδ agonist
  • High selectivity over PPARα and PPARγ receptors (>750-fold and >2500-fold, respectively)
  • Can be used for the study of primary biliary cholangitis