Learn More
Medchemexpress LLC HY-19522A 5mg Medchemexpress, Seladelpar sodium salt CAS: Purity:>98%
SDP
Catalog No. 501932837
Encompass_Preferred
Medchemexpress, HY-19522A 5mg Seladelpar sodium salt CAS: Seladelpar sodium salt (MBX-8025) is an orally active, potent and specific PPARδ agonist with an EC 50 of 2 nM, showing more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2837 Supplier Medchemexpress LLC Supplier No. HY19522A5MG
Medchemexpress, HY-19522A 5mg Seladelpar sodium salt CAS: Seladelpar sodium salt (MBX-8025) is an orally active, potent and specific PPARδ agonist with an EC 50 of 2 nM, showing more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.