Learn More
Medchemexpress LLC Seladelpar sodium salt | 3026272-26-1 | 98.6% | 466.45 | 1 ML
SDP
Catalog No. 5000401134
Encompass_Preferred
Seladelpar sodium salt is an orally active, potent and specific PPARδ agonist with an EC₅₀ of 2 nM. It shows more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors. It is being developed as a lipid-altering agent and can be used for the study of primary biliary cholangitis.
  • Orally active
  • Potent and specific PPARδ agonist (EC₅₀ of 2 nM)
  • More than 750-fold selectivity over PPARα receptors
  • More than 2500-fold selectivity over PPARγ receptors
  • Can be used for the study of primary biliary cholangitis
  • Developed as a lipid-altering agent
Catalog No. 50-004-01134 Supplier Medchemexpress LLC Supplier No. HY19522A10MM/1ML
Seladelpar sodium salt is an orally active, potent and specific PPARδ agonist with an EC₅₀ of 2 nM. It shows more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors. It is being developed as a lipid-altering agent and can be used for the study of primary biliary cholangitis.
  • Orally active
  • Potent and specific PPARδ agonist (EC₅₀ of 2 nM)
  • More than 750-fold selectivity over PPARα receptors
  • More than 2500-fold selectivity over PPARγ receptors
  • Can be used for the study of primary biliary cholangitis
  • Developed as a lipid-altering agent