Learn More
Medchemexpress LLC Selamectin 100mg | 220119-17-5 | 100MG
SDP
Catalog No. 5000239866
Encompass_Preferred
Selamectin a semi-synthetic macrocyclic lactone is a potent parasiticide and anthelminthic Selamectin activates glutamate-gated chloride channels in neurons and pharyngeal muscles to prevent heartworm Lymphatic filariae and nematode infection Selamectin is also a potent P-glycoprotein substrate and a P-glycoprotein inhibitor with an IC50 of 120 nM[1 [2
Catalog No. 50-002-39866 Supplier Medchemexpress LLC Supplier No. HY107212100MG
Selamectin a semi-synthetic macrocyclic lactone is a potent parasiticide and anthelminthic Selamectin activates glutamate-gated chloride channels in neurons and pharyngeal muscles to prevent heartworm Lymphatic filariae and nematode infection Selamectin is also a potent P-glycoprotein substrate and a P-glycoprotein inhibitor with an IC50 of 120 nM[1 [2