Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selamectin | 220119-17-5 | MFCD31621085 | 99.3% | 769.96 g/mol | C43H63NO11 | 5 MG
SDP

Catalog No. 501968371
Encompass_Preferred

Selamectin is a semi-synthetic macrocyclic lactone parasiticide used in research to target nematodes and ectoparasites by activating glutamate-gated chloride channels. Supplied as a small research pack, it is characterized by a defined molecular formula and molecular weight, and is formulated for solubility in DMSO and other vehicles; consult the manufacturer's datasheet for detailed handling and storage guidance.

  • Semi-synthetic macrocyclic lactone parasiticide.
  • Activates glutamate-gated chloride channels in parasites.
  • Effective against nematodes and certain ectoparasites.
  • Molecular formula C43H63NO11 and molecular weight 769.96 g/mol.
  • High purity suitable for research (99.3%).
  • Soluble in DMSO at ≥ 100 mg/mL for in vitro use.
  • Recommended storage: powder frozen for long-term stability.

Catalog No. 50-196-8371 Supplier Medchemexpress LLC Supplier No. HY1072125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selamectin is a semi-synthetic macrocyclic lactone parasiticide used in research to target nematodes and ectoparasites by activating glutamate-gated chloride channels. Supplied as a small research pack, it is characterized by a defined molecular formula and molecular weight, and is formulated for solubility in DMSO and other vehicles; consult the manufacturer's datasheet for detailed handling and storage guidance.

  • Semi-synthetic macrocyclic lactone parasiticide.
  • Activates glutamate-gated chloride channels in parasites.
  • Effective against nematodes and certain ectoparasites.
  • Molecular formula C43H63NO11 and molecular weight 769.96 g/mol.
  • High purity suitable for research (99.3%).
  • Soluble in DMSO at ≥ 100 mg/mL for in vitro use.
  • Recommended storage: powder frozen for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.