Learn More
Med Vet International Selarid For Cats, 5.1 To 15 Pounds, Blue Label, 6 Dose
Catalog No. 502828512
Encompass_Preferred
Selarid (selamectin) kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Selarid is also indicated for the treatment and control of roundworm (Toxocara cati) and intestinal hookworm (Ancylostoma tubaeforme) infections in cats. For use in cats eight weeks of age and older.
Catalog No. 50-282-8512 Supplier Med Vet International Supplier No. RXSELARID6BLV
Selarid (selamectin) kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Selarid is also indicated for the treatment and control of roundworm (Toxocara cati) and intestinal hookworm (Ancylostoma tubaeforme) infections in cats. For use in cats eight weeks of age and older.