Learn More
Med Vet International Selarid For Dogs, 5.1 To 10 Pounds, Purple Label, 6 Dose
Catalog No. 502828085
Encompass_Preferred
Selarid kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Selarid also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of tick infestations due to Dermacentor variabilis in dogs. For use in dogs six weeks of age or older.
Catalog No. 50-282-8085 Supplier Med Vet International Supplier No. RXSELARI6PRV
Selarid kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Selarid also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of tick infestations due to Dermacentor variabilis in dogs. For use in dogs six weeks of age or older.