Learn More
Analytical Sales & Services Select-A-Vial™, 1.0mL Silanized Complete System
Catalog No. 5000807067
Encompass
96 Position Select A Vial - Complete System Racked 1.0mL Silanized Clear Conical Glass Inserts in Rack and Base Tray
Catalog No. 50-008-07067 Supplier Analytical Sales & Services Supplier No. 27001S
96 Position Select A Vial - Complete System Racked 1.0mL Silanized Clear Conical Glass Inserts in Rack and Base Tray